Learn More
Cayman Chemical N2-Isobutyryl Guanosine, Purity: ≥95%, CAS: 64350-24-9, 10 g
SDP
Catalog No. 502942260
Encompass_Preferred
A derivative of guanosine; a precursor in the synthesis of hydrophobic dinucleotide cap analogs used to purify capped mRNA; a precursor in the synthesis of photocaged 3’-S-RNAs used in the study of ribozyme-catalyzed reactions
Catalog No. 50-294-2260 Supplier Cayman Chemical Supplier No. 4028310 G
A derivative of guanosine; a precursor in the synthesis of hydrophobic dinucleotide cap analogs used to purify capped mRNA; a precursor in the synthesis of photocaged 3’-S-RNAs used in the study of ribozyme-catalyzed reactions