Learn More
Medchemexpress LLC N2-Methylguanosine | 2140-77-4 | 98.6% | 297.27 | 100 MG
SDP
Catalog No. 5000439143
Encompass_Preferred
N2-Methylguanosine is a modified nucleoside found in rRNA and tRNA, with specific distributions in both *E. coli* rRNA and eukaryotic tRNA. It can be found in urine and affects the structure and stability of RNA. It is intended for research use only.
  • Modified nucleoside
  • Found in rRNA and tRNA
  • Specific distributions in E. coli rRNA and eukaryotic tRNA
  • Affects RNA structure and stability
  • Suitable for research applications
Catalog No. 50-004-39143 Supplier Medchemexpress LLC Supplier No. HY111647100MG
N2-Methylguanosine is a modified nucleoside found in rRNA and tRNA, with specific distributions in both *E. coli* rRNA and eukaryotic tRNA. It can be found in urine and affects the structure and stability of RNA. It is intended for research use only.
  • Modified nucleoside
  • Found in rRNA and tRNA
  • Specific distributions in E. coli rRNA and eukaryotic tRNA
  • Affects RNA structure and stability
  • Suitable for research applications