Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2-Methylguanosine | 2140-77-4 | 98.6% | 297.27 | 25 MG
SDP

Catalog No. 5000439555
Encompass_Preferred

N2-Methylguanosine is a modified nucleoside found in rRNA and tRNA, with specific distributions in both E. coli rRNA and eukaryotic tRNA. It can also be found in urine and influences the structure and stability of RNA.

  • Forms a stable complex with C, and changes in its conformation and stacking may be critical for tRNA recognizing synthetases.
  • Can alter the base-pairing behavior of RNA and affect the stability of RNA duplexes and hairpin structures. Its mechanism involves steric hindrance, changes in hydrogen bonds, and alterations in the hydration pattern.
  • Molecular weight: 297.27
  • Formula: C11H15N5O5
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Eukaryotes and archaea.

Catalog No. 50-004-39555 Supplier Medchemexpress LLC Supplier No. 25MGHY111647
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-Methylguanosine is a modified nucleoside found in rRNA and tRNA, with specific distributions in both E. coli rRNA and eukaryotic tRNA. It can also be found in urine and influences the structure and stability of RNA.

  • Forms a stable complex with C, and changes in its conformation and stacking may be critical for tRNA recognizing synthetases.
  • Can alter the base-pairing behavior of RNA and affect the stability of RNA duplexes and hairpin structures. Its mechanism involves steric hindrance, changes in hydrogen bonds, and alterations in the hydration pattern.
  • Molecular weight: 297.27
  • Formula: C11H15N5O5
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Eukaryotes and archaea.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.