Learn More
Medchemexpress LLC N2-Methylguanosine | 2140-77-4 | 98.6% | 297.27 | 50 MG
SDP
Catalog No. 5000439144
Encompass_Preferred
N2-Methylguanosine is a commonly modified nucleoside found in rRNA and tRNA, with specific distributions in both E. coli rRNA and eukaryotic tRNA. It can also be found in urine and affects the structure and stability of RNA. It can alter the base-pairing behavior of RNA and affect the stability of RNA duplexes and hairpin structures.
  • Can form a stable complex with C
  • Changes in conformation and stacking may play a crucial role in tRNA recognizing synthetases
  • Mechanism of action involves steric hindrance, changes in hydrogen bonds, and alterations in the hydration pattern
Catalog No. 50-004-39144 Supplier Medchemexpress LLC Supplier No. HY11164750MG
N2-Methylguanosine is a commonly modified nucleoside found in rRNA and tRNA, with specific distributions in both E. coli rRNA and eukaryotic tRNA. It can also be found in urine and affects the structure and stability of RNA. It can alter the base-pairing behavior of RNA and affect the stability of RNA duplexes and hairpin structures.
  • Can form a stable complex with C
  • Changes in conformation and stacking may play a crucial role in tRNA recognizing synthetases
  • Mechanism of action involves steric hindrance, changes in hydrogen bonds, and alterations in the hydration pattern