Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2-methylguanosine | 2140-77-4 | MFCD00057039 | 98.1% | 297.27 g/mol | C11H15N5O5 | 5 MG
SDP

Catalog No. 5000263402
Encompass_Preferred

N2-methylguanosine is a methylated nucleoside present in rRNA and tRNA, supplied as a research-grade analytical standard in powder form. It is intended for biochemical and analytical applications and can influence RNA structure and stability.

  • Provided as a powder for research use only.
  • High purity (~98.1%).
  • Molecular formula C11H15N5O5; molecular weight 297.27 g/mol.
  • Soluble in DMSO (≈7.69 mg/mL with ultrasonic).
  • Recommended storage -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes suitable for analytical use.

Catalog No. 50-002-63402 Supplier Medchemexpress LLC Supplier No. HY1116475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2-methylguanosine is a methylated nucleoside present in rRNA and tRNA, supplied as a research-grade analytical standard in powder form. It is intended for biochemical and analytical applications and can influence RNA structure and stability.

  • Provided as a powder for research use only.
  • High purity (~98.1%).
  • Molecular formula C11H15N5O5; molecular weight 297.27 g/mol.
  • Soluble in DMSO (≈7.69 mg/mL with ultrasonic).
  • Recommended storage -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.