Learn More
Medchemexpress LLC N2,N2-Dimethylguanosine | 2140-67-2 | 99.4% | 311.29 | 1 MG
SDP
Catalog No. 5000437749
Encompass_Preferred
N2,N2-Dimethylguanosine is a methylated nucleoside found in RNA and tRNA. It interferes with reverse transcriptase-mediated cDNA synthesis and impacts high-throughput RNA sequencing efficiency. The compound can be selectively demethylated by AlkB mutant enzymes into N2-methylguanosine, which helps to reduce reverse transcription hindrance.
  • Methylated nucleoside present in RNA and tRNA.
  • Hinders reverse transcriptase-mediated cDNA synthesis.
  • Affects sequencing efficiency in high-throughput RNA sequencing.
  • Can be converted to N2-methylguanosine by AlkB mutant enzymes to reduce reverse transcription hindrance.
Catalog No. 50-004-37749 Supplier Medchemexpress LLC Supplier No. HY1131371MG
N2,N2-Dimethylguanosine is a methylated nucleoside found in RNA and tRNA. It interferes with reverse transcriptase-mediated cDNA synthesis and impacts high-throughput RNA sequencing efficiency. The compound can be selectively demethylated by AlkB mutant enzymes into N2-methylguanosine, which helps to reduce reverse transcription hindrance.
  • Methylated nucleoside present in RNA and tRNA.
  • Hinders reverse transcriptase-mediated cDNA synthesis.
  • Affects sequencing efficiency in high-throughput RNA sequencing.
  • Can be converted to N2-methylguanosine by AlkB mutant enzymes to reduce reverse transcription hindrance.