Learn More
Medchemexpress LLC N2,N2-Dimethylguanosine | 2140-67-2 | 99.4% | 311.29 | 50 MG
SDP
Catalog No. 5000435668
Encompass_Preferred
N2,N2-Dimethylguanosine is a methylated nucleoside present in RNA and a structural modification component of tRNA. It hinders reverse transcriptase-mediated cDNA synthesis and is a key modification affecting sequencing efficiency in high-throughput RNA sequencing. It can be selectively demethylated into N2-methylguanosine by AlkB mutant enzymes (such as D135S/L118V), reducing hindrance to reverse transcription.
  • Methylated nucleoside present in RNA and a structural modification component of tRNA.
  • Hinders reverse transcriptase-mediated cDNA synthesis.
  • Key modification affecting sequencing efficiency in high-throughput RNA sequencing.
  • Can be converted to N2-methylguanosine by AlkB mutant enzymes to reduce hindrance to reverse transcription.
  • Appears as a white to off-white solid.
  • Initial source: Endogenous metabolite.
Catalog No. 50-004-35668 Supplier Medchemexpress LLC Supplier No. HY11313750MG
N2,N2-Dimethylguanosine is a methylated nucleoside present in RNA and a structural modification component of tRNA. It hinders reverse transcriptase-mediated cDNA synthesis and is a key modification affecting sequencing efficiency in high-throughput RNA sequencing. It can be selectively demethylated into N2-methylguanosine by AlkB mutant enzymes (such as D135S/L118V), reducing hindrance to reverse transcription.
  • Methylated nucleoside present in RNA and a structural modification component of tRNA.
  • Hinders reverse transcriptase-mediated cDNA synthesis.
  • Key modification affecting sequencing efficiency in high-throughput RNA sequencing.
  • Can be converted to N2-methylguanosine by AlkB mutant enzymes to reduce hindrance to reverse transcription.
  • Appears as a white to off-white solid.
  • Initial source: Endogenous metabolite.