Learn More
Cayman Chemical N2 N2-DImethylgunosIn 10mg
SDP
Catalog No. 502909987
Encompass_Preferred
A methylated purine nucleoside; formed during the degradation of tRNA; serum levels are elevated in patients with AMML or large cell lung carcinoma
Catalog No. 50-290-9987 Supplier Cayman Chemical Supplier No. 3534710 MG
A methylated purine nucleoside; formed during the degradation of tRNA; serum levels are elevated in patients with AMML or large cell lung carcinoma