Learn More
Cayman Chemical N2,N2-Dimethylguanosine, Purity: ≥95%, CAS: 2140-67-2, 50 mg
SDP
Catalog No. 502909412
Encompass_Preferred
A methylated purine nucleoside; formed during the degradation of tRNA; serum levels are elevated in patients with AMML or large cell lung carcinoma
Catalog No. 50-290-9412 Supplier Cayman Chemical Supplier No. 3534750 MG
A methylated purine nucleoside; formed during the degradation of tRNA; serum levels are elevated in patients with AMML or large cell lung carcinoma