Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine | 75932-02-4 | MFCD00270743 | 95.7% | 590.66 g/mol | C36H34N2O6 | 10 G
SDP

Catalog No. 5000210094
Encompass_Preferred

N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-lysine derivative supplied as a white to off-white solid for research use. It is used as a protected amino acid in peptide synthesis and as a synthetic intermediate in organic chemistry.

  • CAS number: 75932-02-4
  • Chemical formula: C36H34N2O6
  • Molecular weight: 590.66 g/mol
  • Purity: 95.71%
  • Physical form: white to off-white solid
  • Pack sizes: 5 g, 10 g, 25 g, 100 g, larger quantities available
  • Applications: protected amino acid for peptide synthesis and synthetic chemistry
  • For research use only

Catalog No. 50-002-10094 Supplier Medchemexpress LLC Supplier No. HYW04869110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-lysine derivative supplied as a white to off-white solid for research use. It is used as a protected amino acid in peptide synthesis and as a synthetic intermediate in organic chemistry.

  • CAS number: 75932-02-4
  • Chemical formula: C36H34N2O6
  • Molecular weight: 590.66 g/mol
  • Purity: 95.71%
  • Physical form: white to off-white solid
  • Pack sizes: 5 g, 10 g, 25 g, 100 g, larger quantities available
  • Applications: protected amino acid for peptide synthesis and synthetic chemistry
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.