Learn More
Medchemexpress LLC N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine | 75932-02-4 | MFCD00270743 | 95.7% | 590.66 g/mol | C36H34N2O6 | 5 G
SDP
Catalog No. 5000210344
Encompass_Preferred
N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is a bis-Fmoc protected D-lysine derivative used as a protected amino acid building block in peptide synthesis and related organic chemistry research. It provides orthogonal protection on both the α and ε amino groups, allowing selective deprotection and coupling in multi-step syntheses.
  • Bis-Fmoc protection on α and ε amino groups.
  • Used as a protected amino acid in solid-phase peptide synthesis.
  • White to off-white solid; molecular weight 590.66 g/mol.
  • Typical purity around 95.7% by NP-HPLC (batch certificate).
Catalog No. 50-002-10344 Supplier Medchemexpress LLC Supplier No. HYW0486915G
N2,N6-bis(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is a bis-Fmoc protected D-lysine derivative used as a protected amino acid building block in peptide synthesis and related organic chemistry research. It provides orthogonal protection on both the α and ε amino groups, allowing selective deprotection and coupling in multi-step syntheses.
  • Bis-Fmoc protection on α and ε amino groups.
  • Used as a protected amino acid in solid-phase peptide synthesis.
  • White to off-white solid; molecular weight 590.66 g/mol.
  • Typical purity around 95.7% by NP-HPLC (batch certificate).