Learn More
Medchemexpress LLC N2,N6-bis(tert-butoxycarbonyl)-D-lysine | 65360-27-2 | MFCD00076960 | ≥95.0% | 346.42 | C16H30N2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0185285G
Encompass_Preferred
N2,N6-Bis(tert-butoxycarbonyl)-D-lysine is a Boc-protected derivative of D-lysine used as a protected amino acid building block in peptide synthesis and organic synthesis. Protection of both the N2 and N6 amino groups enables selective coupling and deprotection sequences, making it useful for multi-step peptide assembly and derivatization. Supplied as a solid for research use.
Catalog No. 50-003-01870