Learn More
Medchemexpress LLC N2,N6-bis(tert-butoxycarbonyl)-D-lysine | 65360-27-2 | MFCD00076960 | >95.0% | 346.42 g·mol⁻¹ | C16H30N2O6 | 25 G
SDP
Catalog No. 5000303107
Encompass_Preferred
N2,N6-bis(tert-butoxycarbonyl)-D-lysine is a Boc-protected derivative of D-lysine used as a building block in peptide synthesis and organic synthesis. It enables temporary protection of both the α- and ε-amino groups to allow selective functionalization and coupling steps.
  • Provides dual Boc protection for the α- and ε-amino groups of lysine.
  • Facilitates peptide synthesis and selective side-chain modifications.
  • Supplied as a colorless to off-white solid-liquid mixture.
  • High purity (≥95.0%).
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).
  • Available in 5 g, 10 g, 25 g, and 50 g pack sizes.
Catalog No. 50-003-03107 Supplier Medchemexpress LLC Supplier No. 25GHYW018528
N2,N6-bis(tert-butoxycarbonyl)-D-lysine is a Boc-protected derivative of D-lysine used as a building block in peptide synthesis and organic synthesis. It enables temporary protection of both the α- and ε-amino groups to allow selective functionalization and coupling steps.
  • Provides dual Boc protection for the α- and ε-amino groups of lysine.
  • Facilitates peptide synthesis and selective side-chain modifications.
  • Supplied as a colorless to off-white solid-liquid mixture.
  • High purity (≥95.0%).
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).
  • Available in 5 g, 10 g, 25 g, and 50 g pack sizes.