Learn More
Medchemexpress LLC N2,N6,N6,5,7-Pentamethyl-3-(phenylsulfonyl)pyrazolo[1,5-a]pyrimidine-2,6-diamine | 1220646-23-0 | MFCD31544338 | 99.3% | 359.45 g/mol | C17H21N5O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10192425MG
Encompass_Preferred
AVN-492 is a potent, highly selective antagonist of the 5-HT6 (serotonin) receptor used as a research tool in preclinical CNS and cognitive studies. It exhibits picomolar affinity (Ki = 91 pM) and is supplied as a well-characterized solid research compound.
  • High potency: Ki = 91 pM at 5-HT6 receptor.
  • High selectivity for 5-HT6 over related receptors.
  • Research-grade purity (reported ~99.3%).
  • Soluble in DMSO (~50 mg/mL); documented in vivo formulations available.
  • Stable storage recommendations documented for powder and solution forms.
Catalog No. 50-002-48747