Learn More
STEMCELL Technologies N2 Supplement-B, Size: 5 mL
SDP
Catalog No. 501974387
Encompass
N2 Supplement-B, containing iron-poor human apo-transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. N2 Supplement-B can be used in a wide range of protocols to achieve neural induction of human ES/iPS cells and/or downstream differentiation of neural progenitor cells to different neuronal or glial subtypes. N2 Supplement-B is provided as a 100X stock solution.
Catalog No. 50-197-4387 Supplier STEMCELL Technologies Supplier No. 07156
N2 Supplement-B, containing iron-poor human apo-transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. N2 Supplement-B can be used in a wide range of protocols to achieve neural induction of human ES/iPS cells and/or downstream differentiation of neural progenitor cells to different neuronal or glial subtypes. N2 Supplement-B is provided as a 100X stock solution.