Learn More
Sigma Aldrich Fine Chemicals Biosciences N27 A Rat Dopaminergic Neu
Catalog No. 502907133
Encompass
Parkinsons disease is characterized by the death of dopaminergic neurons in the substantia nigra of the brain . In vitro models are critical for understanding the molecular pathology of Parkinsons disease. The N27 immortalized rat dopaminergic cell line has been widely utilized in Parkinsons research for over 20 years. However long-term passaging has led the N27 cell line to become a mixture of cell types with widely variable expression of dopaminergic neural markers such as tyrosine hydroxylase (TH).
Catalog No. 50-290-7133 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCC196
Parkinsons disease is characterized by the death of dopaminergic neurons in the substantia nigra of the brain . In vitro models are critical for understanding the molecular pathology of Parkinsons disease. The N27 immortalized rat dopaminergic cell line has been widely utilized in Parkinsons research for over 20 years. However long-term passaging has led the N27 cell line to become a mixture of cell types with widely variable expression of dopaminergic neural markers such as tyrosine hydroxylase (TH).