Learn More
Cayman Chemical N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine, Purity: ≥95%, CAS: 68892-41-1, 500 mg
SDP
Catalog No. 502991224
Encompass_Preferred
A building block; has been used in the synthesis of oligodeoxyribonucleotides
Catalog No. 50-299-1224 Supplier Cayman Chemical Supplier No. 30472500 MG
A building block; has been used in the synthesis of oligodeoxyribonucleotides