Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N3-C2-NHS ester | 850180-76-6 | MFCD24452814 | 99.8% | 212.16 g/mol | C7H8N4O4 | 10 MG
SDP

Catalog No. 5000201881
Encompass_Preferred

N3-C2-NHS ester is a noncleavable ADC linker and NHS-activated ester reagent used for bioconjugation and the synthesis of antibody-drug conjugates. It is an oil-like, colorless to off-white small molecule with high purity and good solubility in DMSO for research applications.

  • Noncleavable ADC linker for antibody-drug conjugation.
  • NHS-activated ester enables amine-selective bioconjugation.
  • High purity suitable for synthetic chemistry (≈99.8%).
  • Soluble in DMSO; ultrasonic assistance may be required.
  • Store at -20 °C in pure form; follow solvent-storage guidelines.
  • Available in small research sizes, including 10 MG.

Catalog No. 50-002-01881 Supplier Medchemexpress LLC Supplier No. HY12652010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N3-C2-NHS ester is a noncleavable ADC linker and NHS-activated ester reagent used for bioconjugation and the synthesis of antibody-drug conjugates. It is an oil-like, colorless to off-white small molecule with high purity and good solubility in DMSO for research applications.

  • Noncleavable ADC linker for antibody-drug conjugation.
  • NHS-activated ester enables amine-selective bioconjugation.
  • High purity suitable for synthetic chemistry (≈99.8%).
  • Soluble in DMSO; ultrasonic assistance may be required.
  • Store at -20 °C in pure form; follow solvent-storage guidelines.
  • Available in small research sizes, including 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.