Learn More
Medchemexpress LLC N3-C4-NHS ester | 478801-48-8 | 99.5% | 240.22 g/mol | C9H12N4O4 | 250 MG
SDP
Catalog No. 5000202955
Encompass_Preferred
N3-C4-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker and click-chemistry reagent containing an azide group. It is used for bioconjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), and is suitable for ADC synthesis and related chemical biology applications. Purity is 99.53%, molecular weight 240.22 g/mol, formula C9H12N4O4. Storage: pure form -20°C for long-term stability.
  • Noncleavable ADC linker for antibody-drug conjugate synthesis.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO or BCN.
  • High purity: 99.5% to support sensitive conjugation reactions.
  • Stored at -20°C in pure form for up to 3 years.
Catalog No. 50-002-02955 Supplier Medchemexpress LLC Supplier No. HY126522250MG
N3-C4-NHS ester is a noncleavable antibody-drug conjugate (ADC) linker and click-chemistry reagent containing an azide group. It is used for bioconjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), and is suitable for ADC synthesis and related chemical biology applications. Purity is 99.53%, molecular weight 240.22 g/mol, formula C9H12N4O4. Storage: pure form -20°C for long-term stability.
  • Noncleavable ADC linker for antibody-drug conjugate synthesis.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO or BCN.
  • High purity: 99.5% to support sensitive conjugation reactions.
  • Stored at -20°C in pure form for up to 3 years.