Learn More
Medchemexpress LLC N3-Cho bromide (azido-choline bromide) | 2059973-54-3 | 99.6% | 239.11 g/mol | C6H15BrN4O | 5 MG
SDP
Catalog No. 5000284411
Encompass_Preferred
N3-Cho bromide (azido-choline bromide) is an azide-containing click chemistry reagent used to introduce an azido-choline moiety in the synthesis and labeling of cell membrane structures. It is supplied as a small research pack and is intended for use in chemical synthesis and biochemical labeling applications.
  • Contains an azide functional group for click chemistry conjugation.
  • Suitable for synthesis of membrane components and biochemical probes.
  • High purity (≈99.6%) for research applications.
  • Supplied as a 5 mg research pack for small-scale synthesis.
  • Store sealed at low temperature to minimize degradation.
Catalog No. 50-002-84411 Supplier Medchemexpress LLC Supplier No. HYW588725A5MG
N3-Cho bromide (azido-choline bromide) is an azide-containing click chemistry reagent used to introduce an azido-choline moiety in the synthesis and labeling of cell membrane structures. It is supplied as a small research pack and is intended for use in chemical synthesis and biochemical labeling applications.
  • Contains an azide functional group for click chemistry conjugation.
  • Suitable for synthesis of membrane components and biochemical probes.
  • High purity (≈99.6%) for research applications.
  • Supplied as a 5 mg research pack for small-scale synthesis.
  • Store sealed at low temperature to minimize degradation.