Learn More
Medchemexpress LLC N3-cyclopropyl-7-[[4-(1-methylethyl)phenyl]methyl]-7H-pyrrolo[3,2-f]quinazoline-1,3-diamine | 245520-69-8 | MFCD12910520 | 99.7% | 371.48 g·mol⁻¹ | C23H25N5 | 10 MG
SDP
Catalog No. 5000287277
Encompass_Preferred
SCH79797 is a research-grade small molecule antagonist of protease-activated receptor 1 (PAR1). It selectively blocks PAR1-mediated signaling in cellular and preclinical models, reducing thrombin-induced responses and enabling studies of platelet aggregation, vascular signaling, and tissue injury mechanisms.
  • Selective PAR1 antagonism with low-nanomolar potency (IC50 ≈ 70 nM)
  • Inhibits thrombin-induced platelet aggregation in vitro
  • Demonstrated antiproliferative and pro-apoptotic effects in preclinical studies
  • Used in vascular, endothelial, and astrocyte signaling research
  • High chemical purity reported (>99.7%)
  • Intended for laboratory research use only
Catalog No. 50-002-87277 Supplier Medchemexpress LLC Supplier No. HY1499310MG
SCH79797 is a research-grade small molecule antagonist of protease-activated receptor 1 (PAR1). It selectively blocks PAR1-mediated signaling in cellular and preclinical models, reducing thrombin-induced responses and enabling studies of platelet aggregation, vascular signaling, and tissue injury mechanisms.
  • Selective PAR1 antagonism with low-nanomolar potency (IC50 ≈ 70 nM)
  • Inhibits thrombin-induced platelet aggregation in vitro
  • Demonstrated antiproliferative and pro-apoptotic effects in preclinical studies
  • Used in vascular, endothelial, and astrocyte signaling research
  • High chemical purity reported (>99.7%)
  • Intended for laboratory research use only