Learn More
Medchemexpress LLC N3-D-Dap(Fmoc)-OH | 1807631-13-5 | C18H16N4O4 | 250 MG
SDP
Catalog No. 5000374289
Encompass_Preferred
N3-D-Dap(Fmoc)-OH is a click chemistry reagent and an orthogonally protected diaminocarboxylic acid derivative, suitable for use as a component in coupling reactions. It contains an azide group, allowing it to undergo various cycloaddition reactions.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-74289 Supplier Medchemexpress LLC Supplier No. 250MGHY151715
N3-D-Dap(Fmoc)-OH is a click chemistry reagent and an orthogonally protected diaminocarboxylic acid derivative, suitable for use as a component in coupling reactions. It contains an azide group, allowing it to undergo various cycloaddition reactions.
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups