Learn More
Medchemexpress LLC N3-D-Dap(Fmoc)-OH | 1807631-13-5 | C18H16N4O4 | 500 MG
SDP
Catalog No. 5000372177
Encompass_Preferred
N3-D-Dap(Fmoc)-OH is a click chemistry reagent. It can be utilized as a component for coupling via click reaction and as an orthogonally protected diaminocarboxylic acid derivative.
  • Contains an azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-003-72177 Supplier Medchemexpress LLC Supplier No. HY151715500MG
N3-D-Dap(Fmoc)-OH is a click chemistry reagent. It can be utilized as a component for coupling via click reaction and as an orthogonally protected diaminocarboxylic acid derivative.
  • Contains an azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.