Learn More
Medchemexpress LLC N3-L-Val-OH (CHA) | 1217462-63-9 | C11H22N4O2 | 50 MG
SDP
Catalog No. 5000372396
Encompass_Preferred
N3-L-Val-OH (CHA) is a valine derivative containing an azide group, used in click chemistry. It serves as a synthetic intermediate for Valaciclovir (Valacyclovir) and can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Valine derivative
  • Synthetic intermediate for Valaciclovir (Valacyclovir)
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-72396 Supplier Medchemexpress LLC Supplier No. HY15166150MG
N3-L-Val-OH (CHA) is a valine derivative containing an azide group, used in click chemistry. It serves as a synthetic intermediate for Valaciclovir (Valacyclovir) and can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Valine derivative
  • Synthetic intermediate for Valaciclovir (Valacyclovir)
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)