Learn More
Medchemexpress LLC N3-PEG11-CH2CH2Br | 2098982-00-2 | MFCD32696701 | >98.0% | 634.56 g/mol | C24H48BrN3O11 | 25 MG
SDP
Catalog No. 5000283075
Encompass_Preferred
N3-PEG11-CH2CH2Br is a bifunctional PEG11 linker bearing an azide and a terminal bromoethyl group. The hydrophilic PEG11 spacer provides aqueous solubility and flexibility, allowing the molecule to serve as a versatile building block for bioconjugation and probe/linker synthesis in biochemical assays and research workflows.
Catalog No. 50-002-83075 Supplier Medchemexpress LLC Supplier No. HYW19088125MG
N3-PEG11-CH2CH2Br is a bifunctional PEG11 linker bearing an azide and a terminal bromoethyl group. The hydrophilic PEG11 spacer provides aqueous solubility and flexibility, allowing the molecule to serve as a versatile building block for bioconjugation and probe/linker synthesis in biochemical assays and research workflows.