Learn More
Medchemexpress LLC N3-PEG11-CH2CH2Br | 2098982-00-2 | MFCD32696701 | 99.8% | 634.56 g/mol | C24H48BrN3O11 | 50 MG
SDP
Catalog No. 5000282245
Encompass_Preferred
N3-PEG11-CH2CH2Br is a polyethylene glycol (PEG11) linker with an azide (N3) terminus and a bromide-functionalized ethylene spacer. It is a hydrophilic, viscous liquid used in bioconjugation and click-chemistry workflows to improve aqueous solubility and provide a reactive bromide handle for further modification.
  • Azide-terminated PEG11 linker for click chemistry.
  • Bromide-functionalized ethylene spacer provides a reactive handle.
  • Hydrophilic PEG spacer increases solubility in aqueous media.
  • High purity suitable for research applications.
  • Recommended storage conditions maintain stability.
Catalog No. 50-002-82245 Supplier Medchemexpress LLC Supplier No. HYW19088150MG
N3-PEG11-CH2CH2Br is a polyethylene glycol (PEG11) linker with an azide (N3) terminus and a bromide-functionalized ethylene spacer. It is a hydrophilic, viscous liquid used in bioconjugation and click-chemistry workflows to improve aqueous solubility and provide a reactive bromide handle for further modification.
  • Azide-terminated PEG11 linker for click chemistry.
  • Bromide-functionalized ethylene spacer provides a reactive handle.
  • Hydrophilic PEG spacer increases solubility in aqueous media.
  • High purity suitable for research applications.
  • Recommended storage conditions maintain stability.