Learn More
Medchemexpress LLC N3-PEG2-C2-NHS ester | 1312309-64-0 | MFCD20926376 | 97.0% | 300.27 g/mol | C11H16N4O6 | 100 MG
SDP
Catalog No. 5000203615
Encompass_Preferred
N3-PEG2-C2-NHS ester is an azide-functional PEG2 linker supplied as an N-hydroxysuccinimide (NHS) ester for amine coupling and click-chemistry bioconjugation. It is used as a reagent in antibody-drug conjugate (ADC) synthesis and other bioconjugation workflows.
  • Azide functional group enables copper-catalyzed and strain-promoted click chemistry.
  • NHS ester allows rapid conjugation to primary amines.
  • Peg2 spacer improves aqueous solubility and reduces steric hindrance.
  • Non-cleavable two-unit linker suitable for stable bioconjugates.
  • Supplied in small pack sizes for research use, suitable for analytical and preparative labeling.
Catalog No. 50-002-03615 Supplier Medchemexpress LLC Supplier No. HY126526100MG
N3-PEG2-C2-NHS ester is an azide-functional PEG2 linker supplied as an N-hydroxysuccinimide (NHS) ester for amine coupling and click-chemistry bioconjugation. It is used as a reagent in antibody-drug conjugate (ADC) synthesis and other bioconjugation workflows.
  • Azide functional group enables copper-catalyzed and strain-promoted click chemistry.
  • NHS ester allows rapid conjugation to primary amines.
  • Peg2 spacer improves aqueous solubility and reduces steric hindrance.
  • Non-cleavable two-unit linker suitable for stable bioconjugates.
  • Supplied in small pack sizes for research use, suitable for analytical and preparative labeling.