Learn More
Medchemexpress LLC N3-PEG2-C2-NHS ester | 1312309-64-0 | MFCD20926376 | ≥98.0% | 300.27 | C11H16N4O6 | 250 MG
SDP
Catalog No. 5000203737
Encompass_Preferred
N3-PEG2-C2-NHS ester is an azide-functionalized PEG2 NHS ester linker used for bioconjugation and the synthesis of antibody-drug conjugates. The molecule contains an NHS ester for coupling to primary amines and an azide group enabling click chemistry (copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition). It is provided as a high-purity liquid and is soluble in DMSO for research-scale conjugation.
  • Azide-functionalized PEG2 linker for click chemistry
  • NHS ester for efficient amine coupling
  • High purity (≥98.0%) for reliable conjugation
  • Soluble in DMSO (approx. 100 mg/mL) for stock solutions
  • Stable when stored dry at -20°C for extended periods
Catalog No. 50-002-03737 Supplier Medchemexpress LLC Supplier No. HY126526250MG
N3-PEG2-C2-NHS ester is an azide-functionalized PEG2 NHS ester linker used for bioconjugation and the synthesis of antibody-drug conjugates. The molecule contains an NHS ester for coupling to primary amines and an azide group enabling click chemistry (copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition). It is provided as a high-purity liquid and is soluble in DMSO for research-scale conjugation.
  • Azide-functionalized PEG2 linker for click chemistry
  • NHS ester for efficient amine coupling
  • High purity (≥98.0%) for reliable conjugation
  • Soluble in DMSO (approx. 100 mg/mL) for stock solutions
  • Stable when stored dry at -20°C for extended periods