Learn More
Medchemexpress LLC N3-PEG3-C2-NHS ester | 1245718-89-1 | MFCD23726613 | 99.9% | 344.32 | C13H20N4O7 | 100 MG
SDP
Catalog No. 5000203744
Encompass_Preferred
N3-PEG3-C2-NHS ester is a noncleavable, 3-unit polyethylene glycol (PEG) linker bearing an azide group and an N-hydroxysuccinimide (NHS) ester. It is supplied for research use and facilitates bioconjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Noncleavable 3-unit PEG linker for bioconjugation.
  • Contains azide and NHS ester functional groups.
  • Enables CuAAC and SPAAC click chemistry reactions.
  • High purity suitable for research applications.
  • Available in multiple pack sizes including 100 mg.
Catalog No. 50-002-03744 Supplier Medchemexpress LLC Supplier No. HY126528100MG
N3-PEG3-C2-NHS ester is a noncleavable, 3-unit polyethylene glycol (PEG) linker bearing an azide group and an N-hydroxysuccinimide (NHS) ester. It is supplied for research use and facilitates bioconjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Noncleavable 3-unit PEG linker for bioconjugation.
  • Contains azide and NHS ester functional groups.
  • Enables CuAAC and SPAAC click chemistry reactions.
  • High purity suitable for research applications.
  • Available in multiple pack sizes including 100 mg.