Learn More
Medchemexpress LLC N3-peg3-c2-nhs ester | 1245718-89-1 | MFCD23726613 | 99.9% | 344.32 g/mol | C13H20N4O7 | 10 G
SDP
Catalog No. 5000201902
Encompass_Preferred
Azide-terminated PEG3 NHS ester linker for bioconjugation and click-chemistry labeling. The reagent contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is compatible with strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as a high-purity NHS ester for amine conjugation and includes a three-unit PEG spacer to improve solubility and reduce steric hindrance.
  • Azide functional group for click chemistry reactions.
  • NHS ester for efficient conjugation to primary amines.
  • Three-unit PEG spacer to enhance solubility.
  • Noncleavable linker for stable conjugation.
  • High purity suitable for research applications.
  • Available in laboratory-scale 10 g packaging.
Catalog No. 50-002-01902 Supplier Medchemexpress LLC Supplier No. HY12652810G
Azide-terminated PEG3 NHS ester linker for bioconjugation and click-chemistry labeling. The reagent contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is compatible with strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as a high-purity NHS ester for amine conjugation and includes a three-unit PEG spacer to improve solubility and reduce steric hindrance.
  • Azide functional group for click chemistry reactions.
  • NHS ester for efficient conjugation to primary amines.
  • Three-unit PEG spacer to enhance solubility.
  • Noncleavable linker for stable conjugation.
  • High purity suitable for research applications.
  • Available in laboratory-scale 10 g packaging.