Learn More
Medchemexpress LLC N3-peg3-ch2ch2-boc | 252881-73-5 | MFCD22201543 | ≥98.0% | 303.35 g/mol | C13H25N3O5 | 1 G
SDP
Catalog No. 5000202335
Encompass_Preferred
N3-PEG3-CH2CH2-Boc is an azide-functionalized, Boc-protected PEG3 cleavable linker used for bioconjugation and click chemistry applications such as ADC and PROTAC synthesis.
  • Azide functional group enables CuAAC and SPAAC click reactions.
  • Boc-protected terminus allows orthogonal deprotection strategies.
  • Three-unit PEG spacer improves solubility and flexibility in conjugates.
  • High purity suitable for synthesis and analytical workflows.
  • Available in multiple packaging sizes for lab-scale use.
Catalog No. 50-002-02335 Supplier Medchemexpress LLC Supplier No. HY424891G
N3-PEG3-CH2CH2-Boc is an azide-functionalized, Boc-protected PEG3 cleavable linker used for bioconjugation and click chemistry applications such as ADC and PROTAC synthesis.
  • Azide functional group enables CuAAC and SPAAC click reactions.
  • Boc-protected terminus allows orthogonal deprotection strategies.
  • Three-unit PEG spacer improves solubility and flexibility in conjugates.
  • High purity suitable for synthesis and analytical workflows.
  • Available in multiple packaging sizes for lab-scale use.