Learn More
Medchemexpress LLC N3-PEG3-vc-PAB-MMAE | 99.1% | 1352.66 | C67H109N13O16 | 5 MG
SDP
Catalog No. 5000211391
Encompass_Preferred
N3-PEG3-vc-PAB-MMAE is an azide-functionalized drug-linker for antibody-drug conjugate research that combines a PEG3 spacer, a valine-citrulline cleavable peptide with a p-aminobenzyl self-immolative spacer, and the cytotoxic payload monomethyl auristatin E (MMAE). It is designed for bioorthogonal conjugation and ADC construction, supplied as a white to off-white solid with high purity.
  • Azide-functionalized for CuAAC and SPAAC conjugation.
  • Peg3 spacer improves solubility and flexibility of conjugates.
  • Valine-citrulline-PAB linker enables enzymatic release of payload.
  • MMAE payload provides potent antimitotic activity.
  • High molecular weight (1352.66) supports defined stoichiometry in ADCs.
  • High purity (~99.1%) reduces impurities in conjugation reactions.
  • Powder stable at -20°C (3 years) and 4°C (2 years); prepare fresh in solution.
Catalog No. 50-002-11391 Supplier Medchemexpress LLC Supplier No. HY1008745MG
N3-PEG3-vc-PAB-MMAE is an azide-functionalized drug-linker for antibody-drug conjugate research that combines a PEG3 spacer, a valine-citrulline cleavable peptide with a p-aminobenzyl self-immolative spacer, and the cytotoxic payload monomethyl auristatin E (MMAE). It is designed for bioorthogonal conjugation and ADC construction, supplied as a white to off-white solid with high purity.
  • Azide-functionalized for CuAAC and SPAAC conjugation.
  • Peg3 spacer improves solubility and flexibility of conjugates.
  • Valine-citrulline-PAB linker enables enzymatic release of payload.
  • MMAE payload provides potent antimitotic activity.
  • High molecular weight (1352.66) supports defined stoichiometry in ADCs.
  • High purity (~99.1%) reduces impurities in conjugation reactions.
  • Powder stable at -20°C (3 years) and 4°C (2 years); prepare fresh in solution.