Learn More
Medchemexpress LLC N3-PEG3-VC-PAB-MMAF | 2757277-32-8 | 100.0% | 1352.62 | C66H105N13O17 | 25 MG
SDP
Catalog No. 5000204385
Encompass_Preferred
N3-PEG3-VC-PAB-MMAF is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines an azide-functionalized PEG3 spacer, a valine-citrulline (VC) cleavable linker with a para-aminobenzyl (PAB) self-immolative spacer, and the tubulin inhibitor MMAF to enable click chemistry conjugation and controlled payload release.
  • Azide-functionalized PEG3 spacer for click chemistry conjugation.
  • VC-PAB cleavable linker enabling enzymatic release of payload.
  • Includes MMAF payload for tubulin inhibition in ADC constructs.
  • White to off-white solid with high purity suitable for research use.
  • Available in small milligram pack sizes for synthesis workflows.
Catalog No. 50-002-04385 Supplier Medchemexpress LLC Supplier No. HY15373925MG
N3-PEG3-VC-PAB-MMAF is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines an azide-functionalized PEG3 spacer, a valine-citrulline (VC) cleavable linker with a para-aminobenzyl (PAB) self-immolative spacer, and the tubulin inhibitor MMAF to enable click chemistry conjugation and controlled payload release.
  • Azide-functionalized PEG3 spacer for click chemistry conjugation.
  • VC-PAB cleavable linker enabling enzymatic release of payload.
  • Includes MMAF payload for tubulin inhibition in ADC constructs.
  • White to off-white solid with high purity suitable for research use.
  • Available in small milligram pack sizes for synthesis workflows.