Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N3-PEG3-VC-PAB-MMAF | 2757277-32-8 | 100.0% | 1352.62 | C66H105N13O17 | 5 MG
SDP

Catalog No. 5000204486
Encompass_Preferred

N3-PEG3-VC-PAB-MMAF is an azide-functionalized antibody-drug conjugate (ADC) drug-linker conjugate combining a PEG3 spacer, a valine-citrulline cleavable linker with a p-aminobenzyl self-immolative spacer, and the tubulin inhibitor monomethyl auristatin F (MMAF). It is provided as a high-purity click-chemistry reagent for bioconjugation via copper-catalyzed or strain-promoted azide-alkyne cycloaddition.

  • Azide functional group enables CuAAC and SPAAC bioconjugation.
  • Includes valine-citrulline-PAB cleavable linker for intracellular payload release.
  • Contains monomethyl auristatin F as the cytotoxic payload.
  • High purity suitable for research applications (>99.9%).
  • Molecular weight 1352.62; formula C66H105N13O17.
  • Recommended storage at -20°C under inert atmosphere; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04486 Supplier Medchemexpress LLC Supplier No. HY1537395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N3-PEG3-VC-PAB-MMAF is an azide-functionalized antibody-drug conjugate (ADC) drug-linker conjugate combining a PEG3 spacer, a valine-citrulline cleavable linker with a p-aminobenzyl self-immolative spacer, and the tubulin inhibitor monomethyl auristatin F (MMAF). It is provided as a high-purity click-chemistry reagent for bioconjugation via copper-catalyzed or strain-promoted azide-alkyne cycloaddition.

  • Azide functional group enables CuAAC and SPAAC bioconjugation.
  • Includes valine-citrulline-PAB cleavable linker for intracellular payload release.
  • Contains monomethyl auristatin F as the cytotoxic payload.
  • High purity suitable for research applications (>99.9%).
  • Molecular weight 1352.62; formula C66H105N13O17.
  • Recommended storage at -20°C under inert atmosphere; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.