Learn More
Medchemexpress LLC HY-128834 50mg , N3-PEG4-C2-NH2 CAS:951671-92-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12883450MG
Encompass_Preferred
HY-128834 50mg N3-PEG4-C2-NH2 CAS: 951671-92-4 N3-PEG4-C2-NH2 (PROTAC Linker 20) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1739