Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | 98.9% | 250 MG
SDP
Catalog No. 5000342086
Encompass_Preferred
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit PEG linker used in the synthesis of antibody-drug conjugates (ADCs). It acts as a click chemistry reagent, possessing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is intended exclusively for research purposes.
  • Non-cleavable 4-unit PEG linker
  • Used in synthesis of antibody-drug conjugates (ADCs)
  • Functions as a click chemistry reagent with an Azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Intended exclusively for research purposes
Catalog No. 50-003-42086 Supplier Medchemexpress LLC Supplier No. 250MGHY130109
N3-PEG4-C2-NHS ester is a non-cleavable 4-unit PEG linker used in the synthesis of antibody-drug conjugates (ADCs). It acts as a click chemistry reagent, possessing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is intended exclusively for research purposes.
  • Non-cleavable 4-unit PEG linker
  • Used in synthesis of antibody-drug conjugates (ADCs)
  • Functions as a click chemistry reagent with an Azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Intended exclusively for research purposes