Learn More
Medchemexpress LLC N3-PEG4-C2-NHS ester | 944251-24-5 | MFCD13184948 | 98.9% | 388.37 g/mol | C15H24N4O8 | 100 MG
SDP
Catalog No. 5000202176
Encompass_Preferred
N3-PEG4-C2-NHS ester is an azide-terminated polyethylene glycol (PEG4) N-hydroxysuccinimide (NHS) ester used as a bifunctional linker for bioconjugation and click-chemistry applications, including antibody-drug conjugate assembly and labeling of biomolecules.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • NHS ester reacts readily with primary amines to form stable amide bonds.
  • Peg4 spacer provides hydrophilicity and conformational flexibility.
  • High purity supports reliable research-grade conjugation.
  • Stability under recommended cold-storage conditions for long-term use.
Catalog No. 50-002-02176 Supplier Medchemexpress LLC Supplier No. HY130109100MG
N3-PEG4-C2-NHS ester is an azide-terminated polyethylene glycol (PEG4) N-hydroxysuccinimide (NHS) ester used as a bifunctional linker for bioconjugation and click-chemistry applications, including antibody-drug conjugate assembly and labeling of biomolecules.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • NHS ester reacts readily with primary amines to form stable amide bonds.
  • Peg4 spacer provides hydrophilicity and conformational flexibility.
  • High purity supports reliable research-grade conjugation.
  • Stability under recommended cold-storage conditions for long-term use.