Learn More
Cayman Chemical AN3365 (hydrochloride), Purity: ≥98%, CAS: 1234563-16-6, 10 mg
SDP
Catalog No. 502984743
Encompass_Preferred
A LeuRS inhibitor (IC50 = 0.31 µM for the E. coli enzyme); active against a variety of bacteria, including MDR clinical isolates (MICs = 2-8 µg/ml); inhibits protein, RNA, DNA, and fatty acid synthesis in E. coli (IC50s = 8.3, 105, 366, and 396 µg/ml, respectively); reduces the number of CFUs in neutropenic mouse thigh models of E. coli or P. aeruginosa infection at 30 mg/kg
Catalog No. 50-298-4743 Supplier Cayman Chemical Supplier No. 2057210 MG
A LeuRS inhibitor (IC50 = 0.31 µM for the E. coli enzyme); active against a variety of bacteria, including MDR clinical isolates (MICs = 2-8 µg/ml); inhibits protein, RNA, DNA, and fatty acid synthesis in E. coli (IC50s = 8.3, 105, 366, and 396 µg/ml, respectively); reduces the number of CFUs in neutropenic mouse thigh models of E. coli or P. aeruginosa infection at 30 mg/kg