Learn More
Canon Instrument Co N35 Viscosity Standard, 1 L, Hydrocarbon-based, ISO/IEC 17025 Calibration, NIST Traceability, Newtonian liquid
SDP
Catalog No. NC1949570
Encompass_Preferred
N35 Viscosity Standard, 1 L, Hydrocarbon-based, ISO/IEC 17025 Calibration, NIST Traceability, Newtonian liquid
Catalog No. NC1949570 Supplier Canon Instrument Co Supplier No. 9727C37.032
N35 Viscosity Standard, 1 L, Hydrocarbon-based, ISO/IEC 17025 Calibration, NIST Traceability, Newtonian liquid