Learn More
Cayman Chemical N3hydroxybutyrylLHomoserI 10mg
SDP
Supplier:  Cayman Chemical 2769110 MG
Encompass_Preferred
An N-acyl-homoserine lactone produced by the bacterium V. splendidus; has been found in sequencing batch biofilm reactors used in wastewater treatment; racemic version of the V. harveyi autoinducer N-((R)-3-hydroxybutanoyl)-L-homoserine lactone
Catalog No. 50-294-0243