Learn More
Cayman Chemical N3hydroxydecnoylDLHomose 10mg
SDP
Catalog No. 502948469
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; activates SdiA (EC50 = 0.6 µM) in the 14028/pJNS25 strain of S. enterica
Catalog No. 50-294-8469 Supplier Cayman Chemical Supplier No. 3421710 MG
A bacterial quorum-sensing signaling molecule; activates SdiA (EC50 = 0.6 µM) in the 14028/pJNS25 strain of S. enterica