Learn More
Cayman Chemical N3oxobutyrylLHomoserIn 10mg
SDP
Catalog No. 502953224
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; induces expression of rhiI in R. leguminosarum
Catalog No. 50-295-3224 Supplier Cayman Chemical Supplier No. 1302410 MG
A bacterial quorum-sensing signaling molecule; induces expression of rhiI in R. leguminosarum