Learn More
Cayman Chemical N-3-oxo-hexadecanoyl-L-Homoserine lactone, Purity: ≥97%, CAS: 925448-37-9, 10 mg
SDP
Catalog No. 502953037
Encompass_Preferred
An unusual, substituted, long-chain N-acyl-homoserine lactone (AHL) produced by some bacteria
Catalog No. 50-295-3037 Supplier Cayman Chemical Supplier No. 1306210 MG
An unusual, substituted, long-chain N-acyl-homoserine lactone (AHL) produced by some bacteria