Learn More
Cayman Chemical N3oxotetradec7 ZenoylL 10mg
SDP
Catalog No. 502951703
Encompass_Preferred
A bacterial quorum sensing compound that possesses antimicrobial activity, the ability to inhibit biofilm formation, and exhibits immune suppressive activity
Catalog No. 50-295-1703 Supplier Cayman Chemical Supplier No. 1321310 MG
A bacterial quorum sensing compound that possesses antimicrobial activity, the ability to inhibit biofilm formation, and exhibits immune suppressive activity