Learn More
Cayman Chemical N3oxotetradecnoylLHomos 10mg
SDP
Catalog No. 502954158
Encompass_Preferred
A small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism in bacteria; appears later than shorter acyl chain AHLs in developing biofilms; stimulates the production of putisolvin, which in turn, inhibits biofilm formation
Catalog No. 50-295-4158 Supplier Cayman Chemical Supplier No. 1306310 MG
A small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism in bacteria; appears later than shorter acyl chain AHLs in developing biofilms; stimulates the production of putisolvin, which in turn, inhibits biofilm formation