Learn More
Medchemexpress LLC N4-(4-(2-bromo-4,5-difluorophenoxy)phenyl)-L-asparagine hydrochloride | 2450268-84-3 | MFCD28405312 | 99.8% | 451.65 g·mol⁻1 | C16H14BrClF2N2O4 | 100 MG
SDP
Catalog No. 5000239830
Encompass_Preferred
WAY-213613 hydrochloride is the hydrochloride salt of WAY-213613, a potent and selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2). It is provided as a high-purity solid for use in CNS pharmacology, in vitro assays, and in vivo formulation studies, with documented solubility and storage recommendations.
Catalog No. 50-002-39830 Supplier Medchemexpress LLC Supplier No. HY107523A100MG
WAY-213613 hydrochloride is the hydrochloride salt of WAY-213613, a potent and selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2). It is provided as a high-purity solid for use in CNS pharmacology, in vitro assays, and in vivo formulation studies, with documented solubility and storage recommendations.