Learn More
Medchemexpress LLC N4-(4-(2-bromo-4,5-difluorophenoxy)phenyl)-L-asparagine hydrochloride | 2450268-84-3 | MFCD28405312 | 99.8% | 451.65 g/mol | C16H14BrClF2N2O4 | 1 ML
SDP
Catalog No. 5000241450
Encompass_Preferred
WAY-213613 hydrochloride is a potent, selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2) used in central nervous system research. It inhibits EAAT2 with an IC50 of approximately 85 nM and is provided as a ready-to-use solution for pharmacological studies.
  • Potent EAAT2 inhibition (IC50 = 85 nM).
  • Selectivity over EAAT1 and EAAT3.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity (≈99.8%).
  • Suitable for central nervous system research and transporter pharmacology.
Catalog No. 50-002-41450 Supplier Medchemexpress LLC Supplier No. HY107523A10MM 1ML
WAY-213613 hydrochloride is a potent, selective inhibitor of the human excitatory amino acid transporter 2 (EAAT2) used in central nervous system research. It inhibits EAAT2 with an IC50 of approximately 85 nM and is provided as a ready-to-use solution for pharmacological studies.
  • Potent EAAT2 inhibition (IC50 = 85 nM).
  • Selectivity over EAAT1 and EAAT3.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity (≈99.8%).
  • Suitable for central nervous system research and transporter pharmacology.