Learn More
Medchemexpress LLC N4-(4-(dimethylamino)phenyl)-2-(pyridin-4-yl)-5,6,7,8-tetrahydroquinazoline-4,6-diamine | 2230854-93-8 | 98.1% | C21H24N6 | 10MG
SDP
Catalog No. 5000260068
Encompass_Preferred
ARN-21934 is a selective small-molecule inhibitor of human topoisomerase IIα with reported blood-brain barrier penetration and favorable in vivo pharmacokinetics. It inhibits DNA relaxation (IC50 = 2 μM versus etoposide 120 μM) and is supplied for research use only.
  • Selective topoisomerase IIα inhibition (IC50 = 2 μM).
  • Reported blood-brain barrier penetration and favorable in vivo profile.
  • Molecular formula C21H24N6; molecular weight 360.46 g·mol⁻¹.
  • High purity, 98.1%.
  • Available as solid (5-100 mg) and 10 mM solution in DMSO (1 mL).
  • Intended for laboratory research use; not for human use.
Catalog No. 50-002-60068 Supplier Medchemexpress LLC Supplier No. HY13746610MG
ARN-21934 is a selective small-molecule inhibitor of human topoisomerase IIα with reported blood-brain barrier penetration and favorable in vivo pharmacokinetics. It inhibits DNA relaxation (IC50 = 2 μM versus etoposide 120 μM) and is supplied for research use only.
  • Selective topoisomerase IIα inhibition (IC50 = 2 μM).
  • Reported blood-brain barrier penetration and favorable in vivo profile.
  • Molecular formula C21H24N6; molecular weight 360.46 g·mol⁻¹.
  • High purity, 98.1%.
  • Available as solid (5-100 mg) and 10 mM solution in DMSO (1 mL).
  • Intended for laboratory research use; not for human use.