Learn More
Medchemexpress LLC N4-Acetyl-2 -O-methy 5g
SDP
Catalog No. 5000206105
Encompass_Preferred
N4-Acetyl-2 -O-methylcytidine (ac4Cm) is an RNA with dual acetylation and methoxidation modifications found in thermophilic archaea N4-Acetyl-2 -O-methylcytidine has been implicated in the adaptability of archaea in extreme environments enhancing RNA stability through dual modification[1]
Catalog No. 50-002-06105 Supplier Medchemexpress LLC Supplier No. HYW1143275G
N4-Acetyl-2 -O-methylcytidine (ac4Cm) is an RNA with dual acetylation and methoxidation modifications found in thermophilic archaea N4-Acetyl-2 -O-methylcytidine has been implicated in the adaptability of archaea in extreme environments enhancing RNA stability through dual modification[1]