Learn More
AdipoGen N4-Acetylsulfadimethoxine
SDP
Catalog No. 502561763
Encompass_Preferred
Chemical. CAS 555-25-9. Formula C14H16N4O5S. MW 352.37. Synthetic Metabolite of the long-acting sulfonamide antibiotic sulfadimethoxine, used in freshwater aquaculture. Sulfonamide that is benzenesulfonamide substituted by an acetylamino group at position 4 and a 4,6-dimethoxy-pyrimidin-2-yl group at the nitrogen atom. Compound can be used as analytical reference material.
Catalog No. 50-256-1763 Supplier AdipoGen Supplier No. CDXA0298M025
Chemical. CAS 555-25-9. Formula C14H16N4O5S. MW 352.37. Synthetic Metabolite of the long-acting sulfonamide antibiotic sulfadimethoxine, used in freshwater aquaculture. Sulfonamide that is benzenesulfonamide substituted by an acetylamino group at position 4 and a 4,6-dimethoxy-pyrimidin-2-yl group at the nitrogen atom. Compound can be used as analytical reference material.